Learn More
Electron Microscopy Sciences Cryo Storage™ Cane, 50 per pack
SDP
Catalog No. 501902154
Encompass_Preferred
Made from aluminum, 11 5/16" (28.7cm) long. Firmly holds most Cryo Vials for liquid nitrogen storage. It stores up to 6 standard cryo vials.
Catalog No. 50-190-2154 Supplier Electron Microscopy Sciences Supplier No. 6108050
Made from aluminum, 11 5/16" (28.7cm) long. Firmly holds most Cryo Vials for liquid nitrogen storage. It stores up to 6 standard cryo vials.