Learn More
Electron Microscopy Sciences Cryo Storage™ Cane, 10 per pack
SDP
Catalog No. 50996369
Encompass_Preferred
Made from aluminum, 11 5/16" (28.7cm) long. Firmly holds most Cryo Vials for liquid nitrogen storage. It stores up to 6 standard cryo vials.
Catalog No. 50-996-369 Supplier Electron Microscopy Sciences Supplier No. 6108010
Made from aluminum, 11 5/16" (28.7cm) long. Firmly holds most Cryo Vials for liquid nitrogen storage. It stores up to 6 standard cryo vials.