Learn More
Research Products International Corp Cryogenic Storage Rack, Hinged Lid, 50 Tube Capacity, Natural Color, 5 per Case
SDP
Catalog No. 50998022
Encompass
Durable polypropylene storage boxes withstand repeated low temperature storage. Plastic boxes will not deteriorate or saturate like cardboard boxes. Hinged lid provides convenient access to sample tubes. Accommodates all hinged and screw cap 1.5 - 2.0 ml micro-tubes.
Catalog No. 50-998-022 Supplier Research Products International Corp Supplier No. 145633
Durable polypropylene storage boxes withstand repeated low temperature storage. Plastic boxes will not deteriorate or saturate like cardboard boxes. Hinged lid provides convenient access to sample tubes. Accommodates all hinged and screw cap 1.5 - 2.0 ml micro-tubes.