Learn More
Alkali Scientific Cryogenic Storage Box, 100 place, 10 x 10, Blue, PC with Clear Hinged Lid, 5/pk
SDP
Catalog No. 502335854
Encompass_Preferred
Ultra-low temperature Cryogenic Storage Boxes are designed to hold 1.2mL to 2.0mL cryogenic storage vials. An excellent and longlasting replacement for cardboard boxes, they are fully autoclavable and withstand temperatures ranging from -196° to +121°C. They are compatible with all standard 5.25 inch (133mm) freezer racks. Clear lids are hinged and have printed indexing for sample identification purposes and include drainage slots for LN2.
Catalog No. 50-233-5854 Supplier Alkali Scientific Supplier No. MT2200
Ultra-low temperature Cryogenic Storage Boxes are designed to hold 1.2mL to 2.0mL cryogenic storage vials. An excellent and longlasting replacement for cardboard boxes, they are fully autoclavable and withstand temperatures ranging from -196° to +121°C. They are compatible with all standard 5.25 inch (133mm) freezer racks. Clear lids are hinged and have printed indexing for sample identification purposes and include drainage slots for LN2.