Learn More
Andwin Scientific CRYO STRIP LABELS WHITE 1700CS
SDP
Catalog No. NC0416294
Encompass_Preferred
The Cryogenic Dot and Strip Labels can be utilized for all cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications. This product has a unique adhesive that can resist extreme temperatures from -196 to 121C (-321 t
Catalog No. NC0416294 Supplier Andwin Scientific Supplier No. 89097572
The Cryogenic Dot and Strip Labels can be utilized for all cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications. This product has a unique adhesive that can resist extreme temperatures from -196 to 121C (-321 t