Learn More
GA International Transparent Cryogenic Tape for Frozen Surfaces 1" x 50' / 25.4mm x 15m Transparent 1" / 25.4mm From -196°C to +150°C (-321°F to +302°F)
Catalog No. NC1558779
Encompass_Preferred
Transparent CryoSTUCK tape comes in a roll format to identify already frozen plastic containers. The tapes are designed for long-term cryo-preservation in vapor phase liquid nitrogen, ultra-low temperature freezers, and dry ice (-80°C). Our transparent CryoSTUCK TAQ-class tapes are waterproof and suitable for immersion in a water bath.This tape can be conveniently inscribed with our permanent cryogenic markers.
Catalog No. NC1558779 Supplier GA International Supplier No. TAQ25C150
Transparent CryoSTUCK tape comes in a roll format to identify already frozen plastic containers. The tapes are designed for long-term cryo-preservation in vapor phase liquid nitrogen, ultra-low temperature freezers, and dry ice (-80°C). Our transparent CryoSTUCK TAQ-class tapes are waterproof and suitable for immersion in a water bath.This tape can be conveniently inscribed with our permanent cryogenic markers.