Learn More
GA International Cryogenic Linerless Tape for Frozen Surfaces with GatorCUT™ (Patented) 1" x 100' / 25.4mm x 30m Transparent From -196°C to +121°C (-320.8°F to +249.8°F)
Supplier:  GA International TXC25CP3100
Encompass_Preferred
Durable CryoSTUCK tape, designed to stick to frozen surfaces*, including cryo vials boxes, and long-term storage in liquid nitrogen or deep-freeze laboratory freezers without losing its adhesive properties. The cryogenic linerless tape also resists high temperatures, making it a versatile option for professionals in diverse environments. Ideal for use in scientific applications where materials need to be stored at very low or high temperatures from -196°C to +120°C.The tape also features GatorCUT technology, which allows for precise and accurate cutting of the tape to the desired length. This makes it easy to label and identify samples or storage containers in cryogenic environments. Offered in a clear format, this self-wound tape allows for clear sample visualization.Transparent CryoSTUCK tape is a versatile, durable, and reliable cryo tape for the fields of biology, medicine, as well as scientific research and development.* To ensure proper adhesion, please remove excess frost
Catalog No. NC2879381