Learn More
Research Products International Corp Cryogenic Tubes, External Threaded, 2ml, 47.3mm H, 500/cs
SDP
Catalog No. 502252957
Encompass
Cryogenic Tubes are designed for safe storage and transportation of biological materials. Ideal for a variety of applications including boiling, freezing and centrifuging.Tubes feature pre-printed graduations, white marking area, and star foot base to lock into rack for easy one-hand use at workstations. Thick wall polypropylene tubes with silicone O-rings maintain positive seal at temperatures as low as-196°C and up to 121°C and have 12.5mm diameter. Supplied 50 tubes per bag.
Withstand -196°C (vapor)
Printed graduations, conical
Sterile, RNase, DNase and Pyrogen free
This item is not returnable. View return policy
Catalog No. 50-225-2957 Supplier Research Products International Corp Supplier No. 248562
Cryogenic Tubes are designed for safe storage and transportation of biological materials. Ideal for a variety of applications including boiling, freezing and centrifuging.Tubes feature pre-printed graduations, white marking area, and star foot base to lock into rack for easy one-hand use at workstations. Thick wall polypropylene tubes with silicone O-rings maintain positive seal at temperatures as low as-196°C and up to 121°C and have 12.5mm diameter. Supplied 50 tubes per bag.
Withstand -196°C (vapor)
Printed graduations, conical
Sterile, RNase, DNase and Pyrogen free