Learn More
GA International Thermal-Transfer Cryogenic Labels for Frozen Vials & Surfaces with SimPEEL™ Technology (Patent Pending) 1" x 1" / 25.4mm x 25.4mm Rectangle White From -196°C to +121°C (-321°F to +250°F)
Catalog No. NC2740884
Encompass_Preferred
These cryogenic labels provide an all-in-one solution for cryo storage that can firmly adhere to ambient and already frozen* surfaces (as low as -80°C/-112°F) without the need for thawing, cure time, or any other special treatment. With the addition of SimPEEL technology, these labels can now be peeled from their liner quickly and effortlessly via the adhesive-free tab that ensures the labels will not stick to your hands or gloves during application and will maintain excellent long-term adhesion when stored under cryogenic conditions. Due to their unique adhesive and conformable design, these cryo labels for frozen surfaces can be applied onto both flat and curved surfaces, ideal for identifying cryo vials, microtubes, as well as cardboard and polycarbonate storage boxes destined for long-term storage in liquid nitrogen tanks (-196°C/-321°F), ultra-low temperature freezers (-80°C) and dry ice transportation. Ideally suited for identifying vials destined for immediate immersion in liqui
Catalog No. NC2740884 Supplier GA International Supplier No. SPUC29NPC11WH
These cryogenic labels provide an all-in-one solution for cryo storage that can firmly adhere to ambient and already frozen* surfaces (as low as -80°C/-112°F) without the need for thawing, cure time, or any other special treatment. With the addition of SimPEEL technology, these labels can now be peeled from their liner quickly and effortlessly via the adhesive-free tab that ensures the labels will not stick to your hands or gloves during application and will maintain excellent long-term adhesion when stored under cryogenic conditions. Due to their unique adhesive and conformable design, these cryo labels for frozen surfaces can be applied onto both flat and curved surfaces, ideal for identifying cryo vials, microtubes, as well as cardboard and polycarbonate storage boxes destined for long-term storage in liquid nitrogen tanks (-196°C/-321°F), ultra-low temperature freezers (-80°C) and dry ice transportation. Ideally suited for identifying vials destined for immediate immersion in liqui