Learn More
Alkali Scientific ASI™ Cryo Tissue Container, HDPE / PP, 1.5ml, Sterile to SAL10-6, 100/ Cs
SDP
Catalog No. 503029218
Encompass_Preferred
Cryo Tissue Container is suitable for use in -80℃ deep freezer or in the vapor-phased liquid nitrogen up to -178℃. The stackable block-shaped structure is convenient for storing and transporting samples. ▪ Use only in vapor-phased liquid nitrogen▪ Stackable feature▪ Two marking area on outside
Catalog No. 50-302-9218 Supplier Alkali Scientific Supplier No. SM455
Cryo Tissue Container is suitable for use in -80℃ deep freezer or in the vapor-phased liquid nitrogen up to -178℃. The stackable block-shaped structure is convenient for storing and transporting samples. ▪ Use only in vapor-phased liquid nitrogen▪ Stackable feature▪ Two marking area on outside