Learn More
Electron Microscopy Sciences Cryo Transport Box for 10 Tubes
SDP
Catalog No. 50996440
Encompass_Preferred
Transport boxes are ideal for transporting and organizing Cryo Vials. The boxes are made from high impact polystyrene and have a durable construction and are designed to hold 1.0-2.0 ml Cryo Vials. Made from high impact polystyrene. Used to organizes and hold 1.0 - 2.0 ml Cryo Vials.
Catalog No. 50-996-440 Supplier Electron Microscopy Sciences Supplier No. 6200410
Transport boxes are ideal for transporting and organizing Cryo Vials. The boxes are made from high impact polystyrene and have a durable construction and are designed to hold 1.0-2.0 ml Cryo Vials. Made from high impact polystyrene. Used to organizes and hold 1.0 - 2.0 ml Cryo Vials.