Learn More
Electron Microscopy Sciences Cryo Transport Box for 5 Tubes, 20/pk
SDP
Catalog No. 50996439
Encompass_Preferred
Transport boxes are ideal for transporting and organizing Cryo Vials. The boxes are made from high impact polystyrene and have a durable construction and are designed to hold 1.0-2.0 ml Cryo Vials.
Catalog No. 50-996-439 Supplier Electron Microscopy Sciences Supplier No. 6200405
Transport boxes are ideal for transporting and organizing Cryo Vials. The boxes are made from high impact polystyrene and have a durable construction and are designed to hold 1.0-2.0 ml Cryo Vials.