Learn More
MTC Bio Cryogenic Vials, externally threaded caps with silicone O-ring, 5mL, sterile, printed graduations and marking area, self-standing star foot, 50-place locking rack included with each case, 10 resealable bags of 50 tubes, 500/cs
Catalog No. 502268509
Encompass_Preferred
MTC Bio cryogenic vials are designed to hold, store and preserve biological materials for days, weeks, years, or decades. These thick-walled tubes are outfitted with silicone O-rings that allow them to maintain their seal at temperatures as low as -196°C. Sterilized by gamma irradiation, the tubes are packed in reasealable bags of 50 tubes (10 bags per case) and are sterilzied by gamma. The self-standing starfoot base mates perfectly with a FREE included rack so that one hand is all that is need to install and remove caps. RNase, DNase, pyrogen, & endotoxin free
This item is not returnable. View return policy
Catalog No. 50-226-8509 Supplier MTC Bio Supplier No. V3805
MTC Bio cryogenic vials are designed to hold, store and preserve biological materials for days, weeks, years, or decades. These thick-walled tubes are outfitted with silicone O-rings that allow them to maintain their seal at temperatures as low as -196°C. Sterilized by gamma irradiation, the tubes are packed in reasealable bags of 50 tubes (10 bags per case) and are sterilzied by gamma. The self-standing starfoot base mates perfectly with a FREE included rack so that one hand is all that is need to install and remove caps. RNase, DNase, pyrogen, & endotoxin free