Learn More
Research Products International Corp Cryo Tubes & Caps 1.8Ml 500/Pk
SDP
Catalog No. 501537664
Encompass
CryoFreeze Cryogenic Tubes are designed for safe storage and transportation of biological materials. Ideal for a variety of applications including boiling, freezing and centrifuging. Tubes are manufactured from FDA-approved, medical grade polypropylene with HDPE caps. Tubes feature pre-printed black 100μL graduations and a significant white marking area on all tubes. Self standing tubes offer stargrip interlocking base and allow one-hand use in workstations.
Tubes comply with the In Vitro Devices Directive (IVD). Packaged in resealable stand-up pouches that minimize the use of benchtop space.
Withstand -196°C (vapor)
FDA-approved, medical grade
Sterile, RNase, DNase-free
Clearly marked graduations
Catalog No. 50-153-7664 Supplier Research Products International Corp Supplier No. 212604
CryoFreeze Cryogenic Tubes are designed for safe storage and transportation of biological materials. Ideal for a variety of applications including boiling, freezing and centrifuging. Tubes are manufactured from FDA-approved, medical grade polypropylene with HDPE caps. Tubes feature pre-printed black 100μL graduations and a significant white marking area on all tubes. Self standing tubes offer stargrip interlocking base and allow one-hand use in workstations.
Tubes comply with the In Vitro Devices Directive (IVD). Packaged in resealable stand-up pouches that minimize the use of benchtop space.
Withstand -196°C (vapor)
FDA-approved, medical grade
Sterile, RNase, DNase-free
Clearly marked graduations