Learn More
Electron Microscopy Sciences Cryogenic Vial Box with Lid, White, 75 mm, 133 x 133 mm Footprint
SDP
Catalog No. 501926814
Encompass_Preferred
Store cryogenic vials in liquid nitrogen or in mechanical freezers in our freezer storage boxes. Box footprint is 133 x 133 mm. Select a box and a partition that fit the size and number of vials for your needs. Colored cardboard boxes are ideal for identification and sorting purposes.
Color: White
Height: 75 mm
Footprint: 133 x 133 mm
This item is not returnable. View return policy
Catalog No. 50-192-6814 Supplier Electron Microscopy Sciences Supplier No. 41111
Store cryogenic vials in liquid nitrogen or in mechanical freezers in our freezer storage boxes. Box footprint is 133 x 133 mm. Select a box and a partition that fit the size and number of vials for your needs. Colored cardboard boxes are ideal for identification and sorting purposes.
Color: White
Height: 75 mm
Footprint: 133 x 133 mm