Learn More
Electron Microscopy Sciences Cryogenic Vial Canes, 290 mm, 5 Vials
SDP
Catalog No. 501926628
Encompass_Preferred
Aluminum canes hold vials for freezer storage or immersion in LN2 Dewar. For use in liquid phase, use approved vials only. Canes bend to fit all major cryogenic vial brands.
Catalog No. 50-192-6628 Supplier Electron Microscopy Sciences Supplier No. 6106703
Aluminum canes hold vials for freezer storage or immersion in LN2 Dewar. For use in liquid phase, use approved vials only. Canes bend to fit all major cryogenic vial brands.