Learn More
CELLTREAT Scientific Products 100 Place Storage Box for CF Cryogenic Vial, Polycarbonate, Non-sterile
SDP
Catalog No. 501149103
Encompass_Preferred
CELLTREAT Scientific Products Storage Box, Crogenic Vial, 100 Place. CELLTREAT Freeze Cryogenic Vial Storage Boxes are made from durable polycarbonate material designed to withstand a wide temperature range and is a reusable, autoclavable alternative to disposable racks. Ventilation holes in the lids and bases prevent vacuum sealing in freezer. Storage Boxes are labeled with numeric markings on the base of the box for easy identification of individual positions, with or without the lid. 100-place rack features ridges along the sides to improve handling. Fits 1mL - 1.8mL vials. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 5/case.
Catalog No. 50-114-9103 Supplier CELLTREAT Scientific Products Supplier No. 229945
CELLTREAT Scientific Products Storage Box, Crogenic Vial, 100 Place. CELLTREAT Freeze Cryogenic Vial Storage Boxes are made from durable polycarbonate material designed to withstand a wide temperature range and is a reusable, autoclavable alternative to disposable racks. Ventilation holes in the lids and bases prevent vacuum sealing in freezer. Storage Boxes are labeled with numeric markings on the base of the box for easy identification of individual positions, with or without the lid. 100-place rack features ridges along the sides to improve handling. Fits 1mL - 1.8mL vials. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 5/case.