Learn More
CELLTREAT Scientific Products 25 Place Storage Box for CF Cryogenic Vial, Polycarbonate, Non-sterile
SDP
Catalog No. 501149100
Encompass_Preferred
CELLTREAT Scientific Products Storage Box, CF Cryogenic Vial, 25 Place. CELLTREAT Freeze Cryogenic Vial Storage Boxes are made from durable polycarbonate material designed to withstand a wide temperature range and is a reusable, autoclavable alternative to disposable racks. Ventilation holes in the lids and bases prevent vacuum sealing in freezer. Storage Boxes are labeled with numeric markings on the base of the box for easy identification of individual positions, with or without the lid. Fits 1mL - 1.8mL vials. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 5/case.
Catalog No. 50-114-9100 Supplier CELLTREAT Scientific Products Supplier No. 229942
CELLTREAT Scientific Products Storage Box, CF Cryogenic Vial, 25 Place. CELLTREAT Freeze Cryogenic Vial Storage Boxes are made from durable polycarbonate material designed to withstand a wide temperature range and is a reusable, autoclavable alternative to disposable racks. Ventilation holes in the lids and bases prevent vacuum sealing in freezer. Storage Boxes are labeled with numeric markings on the base of the box for easy identification of individual positions, with or without the lid. Fits 1mL - 1.8mL vials. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 5/case.