Learn More
GA International Cryogenic Color Dots Labels 0.75" / 19.1mm Circle White From -196°C to +110°C (-321°F to +230°F)
Catalog No. NC3304392
Encompass_Preferred
These small diameter round cryo labels for tube tops are made with a strong adhesive ideal for -80°C freezers, dry ice, and liquid nitrogen storage*. Specially designed to comfortably fit 15 ml and 50 ml Falcon tube caps, bottle caps, as well as cryo and freezer boxes, these circular, waterproof, cryo dot labels have a permanent adhesive that can withstand prolonged storage inside ultra-low temperature freezers. The freezer dot labels are supplied as Hagaki sheets and are available in 15 individual colors as well as packs of 15 assorted colors for color coding and rapid identification.
Catalog No. NC3304392 Supplier GA International Supplier No. LT19WH
These small diameter round cryo labels for tube tops are made with a strong adhesive ideal for -80°C freezers, dry ice, and liquid nitrogen storage*. Specially designed to comfortably fit 15 ml and 50 ml Falcon tube caps, bottle caps, as well as cryo and freezer boxes, these circular, waterproof, cryo dot labels have a permanent adhesive that can withstand prolonged storage inside ultra-low temperature freezers. The freezer dot labels are supplied as Hagaki sheets and are available in 15 individual colors as well as packs of 15 assorted colors for color coding and rapid identification.