Learn More
GA International Wrap-Around Cryogenic & Autoclave Resistant Self-Laminating Laser Labels 1" x 0.5" + 0.75" (wrap) / 25.4mm x 12.7mm + 19.1mm (wrap) Rectangle White + Transparent From -196°C to +150°C (-321°F to +302°F)
Catalog No. NC1327850
Encompass_Preferred
Cryogenic wrap-around laser labels for long-term cryo-storage in liquid nitrogen and deep freeze harsh environments. Temperature range from -196°C to +150°C. Resistant to autoclave, boiling water, as well as spraying with ethanol, IPA, acetone, and other chemicals and detergents. Cryo labels accept desktop laser printing and permanent marker. Water-resistant; Wear and tear proof; Do not jam printers; Resistant to UV and gamma irradiation.FDA Compliance: FDA 21 CFR 175.105 for indirect food contact.
Catalog No. NC1327850 Supplier GA International Supplier No. DFSL13WH
Cryogenic wrap-around laser labels for long-term cryo-storage in liquid nitrogen and deep freeze harsh environments. Temperature range from -196°C to +150°C. Resistant to autoclave, boiling water, as well as spraying with ethanol, IPA, acetone, and other chemicals and detergents. Cryo labels accept desktop laser printing and permanent marker. Water-resistant; Wear and tear proof; Do not jam printers; Resistant to UV and gamma irradiation.FDA Compliance: FDA 21 CFR 175.105 for indirect food contact.