Learn More
CELLULAR TECHNOLOGY LTD CRYOABC FREEZING MEDIUM KIT
SDP
Catalog No. NC9711917
Encompass
Ctl-Cryo Abc Media Kit. 10Ml (3 Vials Per Kit, Mixing Required). Recommended For 100 Million Cells. The Ctl-Cryo Abc Kit Is A Serum-Free Freezing Media Kit Formulated To Cryopreserve Freshly-Isolated Pbmc. When Adhering To Ctl'S Protocols, Ctl-Cryo Media Ensures A Higher Then 90% Viability (Typically >95%) Of Pbmc After Thawing, And A Recovery Rate Of Greater Then 70% (Up To 95%).
Catalog No. NC9711917 Supplier CELLULAR TECHNOLOGY LTD Supplier No. CTLCABC
Ctl-Cryo Abc Media Kit. 10Ml (3 Vials Per Kit, Mixing Required). Recommended For 100 Million Cells. The Ctl-Cryo Abc Kit Is A Serum-Free Freezing Media Kit Formulated To Cryopreserve Freshly-Isolated Pbmc. When Adhering To Ctl'S Protocols, Ctl-Cryo Media Ensures A Higher Then 90% Viability (Typically >95%) Of Pbmc After Thawing, And A Recovery Rate Of Greater Then 70% (Up To 95%).