Learn More
Research Products International Corp Cryo-Babies, 1.50"" x 0.50"", YLW, 750/rl
SDP
Catalog No. 502229586
Encompass
Cryogenic labels for direct thermal printers. Direct Thermal Cryo-Tags are formatted to work quickly and easily with the Dymo Labelwriter and most direct thermal printers. Quickly label your samples with sharp text, clear barcodes or a combination of both! Cryogenic labels are suitable for a variety of applications.
Recommended Temperature: -196°C to +70°C
Catalog No. 50-222-9586 Supplier Research Products International Corp Supplier No. 249401Y
Cryogenic labels for direct thermal printers. Direct Thermal Cryo-Tags are formatted to work quickly and easily with the Dymo Labelwriter and most direct thermal printers. Quickly label your samples with sharp text, clear barcodes or a combination of both! Cryogenic labels are suitable for a variety of applications.
Recommended Temperature: -196°C to +70°C