Learn More
Research Products International Corp Cryo-Babies Labels for 0.5ml Tubes, 0.94 x 0.5 Inches, White, 1,000 per Roll
SDP
Catalog No. 501367708
Encompass
Cryogenic labels (Cryo-Babies) are flexible polyolefin labels that withstand conventional and cryogenic storage conditions (vapor and liquid phase nitrogen storage at -196°C). The labels are chemically inert with a solvent resistant acrylic adhesive that will adhere to all plastics, glass and metal.
Catalog No. 50-136-7708 Supplier Research Products International Corp Supplier No. 247180
Cryogenic labels (Cryo-Babies) are flexible polyolefin labels that withstand conventional and cryogenic storage conditions (vapor and liquid phase nitrogen storage at -196°C). The labels are chemically inert with a solvent resistant acrylic adhesive that will adhere to all plastics, glass and metal.