Learn More
USA Scientific Inc Cryo-Babies 1.28 x 0.5" orange
Catalog No. 502744851
Encompass
Cryo-Babies, large, orange labels for 1.5 mL and 2.0 mL tubes, 1.28 inch L x 0.5 inch W, 1000/roll. For -196C to +150C.
Catalog No. 50-274-4851 Supplier USA Scientific Inc Supplier No. 91882003
Cryo-Babies, large, orange labels for 1.5 mL and 2.0 mL tubes, 1.28 inch L x 0.5 inch W, 1000/roll. For -196C to +150C.