Learn More
USA Scientific Inc Cryo-Babies, 1.28 x .5", red,
Catalog No. 502744859
Encompass
Cryo-Babies, large, red labels for 1.5 mL and 2.0 mL tubes, 1.28 inch L x 0.5 inch W, 1000/roll. For -196C to +150C.
Catalog No. 50-274-4859 Supplier USA Scientific Inc Supplier No. 91882004
Cryo-Babies, large, red labels for 1.5 mL and 2.0 mL tubes, 1.28 inch L x 0.5 inch W, 1000/roll. For -196C to +150C.