Learn More
Electron Microscopy Sciences Cryo-Tag Thermal Transfer Labels, 20.6 x 7 mm
SDP
Supplier:  Electron Microscopy Sciences 7759265
Encompass_Preferred
Thermal Transfer Cryo-Tags are pre-cut, peel-off labels that are designed for use in thermal transfer printers. The labels are a reliable and convenient way to identify samples with text or a barcode on microtubes, microplates, bottles and other laboratory containers. The labels are resistent to degradation from scuffing, chemicals, moisture and wide temperature fluctuation. The unique solvent-resistant, acrylic-based adhesive provides excellent cold temperature performance. Specially coated for use with a resin ribbon that provides the best performance under extreme conditions.
Size: 20.6 x 7 mm
Use: 0.2ml PCR tubes
Catalog No. 50-190-2262