Learn More
Research Products International Corp Cryo-Babies Labels, Laser Sheet, 1.5 - 2.0ml Tubes, 1.28 x 0.5 Inches, Green, 1,700 per Package
SDP
Supplier:  Research Products International Corp 247127G
Encompass
Cryo-Babies are designed to offer superior temperature and chemical resistance. Durable labels withstand cryogenic liquid nitrogen storage, boiling water baths, organic solvents, caustic agents and other laboratory chemicals. Cryo-Babies fit cryogenic vials and 1.5 - 2.0 ml micro-tubes. Chemically inert poly vinyl labels adhere to all polypropylene and polyethylene surfaces without cracking, peeling or degrading and accept any marking instrument.
Catalog No. 50-476-408