Learn More
Research Products International Corp Cryo-Babies Labels/Tough-Spot Set for Laser Printers, Fits 1.5ml Tubes, 1360 Sets per Package
SDP
Catalog No. 50550265
Encompass
Cryo-Babies and Tough-Spot combo set includes both a 1 1/4 x 1/2 inch side label and a 3/8 inch diameter top label on a white laser sheet. Ideal format and size for 1.5-2.0ml micro-tubes and cryogenic vials. Simplifies labeling and allows faster identification of tubes. Flexible labels are temperature and chemical resistant. Label sheets are specially designed for use with laser printers.
Catalog No. 50-550-265 Supplier Research Products International Corp Supplier No. 247170
Cryo-Babies and Tough-Spot combo set includes both a 1 1/4 x 1/2 inch side label and a 3/8 inch diameter top label on a white laser sheet. Ideal format and size for 1.5-2.0ml micro-tubes and cryogenic vials. Simplifies labeling and allows faster identification of tubes. Flexible labels are temperature and chemical resistant. Label sheets are specially designed for use with laser printers.