Learn More
Labsource Inc Diversified Biotech LCRY-2380-G Laser Cryo-Babies, 0.94 x 0.50", 2,380/pk - Green
SDP
Catalog No. NC9551719
Encompass
This item must be purchased in multiples of 5 rolls per supplier requirements. Our label sheets are specially designed to permanently accept laser printing. Our patented Cryo-Babies and Cryo-Tags withstand both long term cryogenic storage conditions and conventional freezer storage. These labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic vials, microtubes, test tubes, glass vials, beakers and metal racks. Cryo-Babies and Cryo-Tags also withstand flash/snap freezing in liquid nitrogen, boiling water baths at 100°C and dry heat up to 150°C. Perfect for easy sample identification and storage!
Catalog No. NC9551719 Supplier Labsource Inc Supplier No. L60345PK
This item must be purchased in multiples of 5 rolls per supplier requirements. Our label sheets are specially designed to permanently accept laser printing. Our patented Cryo-Babies and Cryo-Tags withstand both long term cryogenic storage conditions and conventional freezer storage. These labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic vials, microtubes, test tubes, glass vials, beakers and metal racks. Cryo-Babies and Cryo-Tags also withstand flash/snap freezing in liquid nitrogen, boiling water baths at 100°C and dry heat up to 150°C. Perfect for easy sample identification and storage!