Learn More
Andwin Scientific CRYOBABIES RBW 24X13 2380/PK
SDP
Catalog No. 50727677
Encompass_Preferred
Suitable for laser printing, the 24X13 Cryo-Babies stay-flat labels in Rainbow can be used on 1.5mL tubes. Made of polyolefin and acrylic adhesive, it is chemical and temperature resistant from -196-150 deg.C. It comes in 2380 a pack.
Catalog No. 50-727-677 Supplier Andwin Scientific Supplier No. 7756105
Suitable for laser printing, the 24X13 Cryo-Babies stay-flat labels in Rainbow can be used on 1.5mL tubes. Made of polyolefin and acrylic adhesive, it is chemical and temperature resistant from -196-150 deg.C. It comes in 2380 a pack.