Learn More
Andwin Scientific CRYOBABIES WH 24X13 2380/PK
SDP
Catalog No. 50727678
Encompass_Preferred
The 0.94" L x 0.5" W Cryo-Babies labels can withstand most organic solvents, caustic agents, temperatures from -196C to 100C. This product can still be utilized while under oven, autoclaving, or extreme freezer temperatures. The labels can also be used on several surfaces like plastics, glass, and metals without cracking, peeling, or degrading. This product is sold in the color of white.Ideal Use: 0.5 ml tubes
Catalog No. 50-727-678 Supplier Andwin Scientific Supplier No. 77561W
The 0.94" L x 0.5" W Cryo-Babies labels can withstand most organic solvents, caustic agents, temperatures from -196C to 100C. This product can still be utilized while under oven, autoclaving, or extreme freezer temperatures. The labels can also be used on several surfaces like plastics, glass, and metals without cracking, peeling, or degrading. This product is sold in the color of white.Ideal Use: 0.5 ml tubes