Learn More
Andwin Scientific CRYOBABIES YL 24X13 2380/PK
SDP
Catalog No. 50727679
Encompass_Preferred
Suitable for laser printing, the 24X13 Cryo-Babies stay-flat labels in Yellow can be used on 1.5mL tubes. Made of polyolefin and acrylic adhesive, it is chemical and temperature resistant from -196-150 deg.C. It comes in 2380 a pack.
Catalog No. 50-727-679 Supplier Andwin Scientific Supplier No. 77561Y
Suitable for laser printing, the 24X13 Cryo-Babies stay-flat labels in Yellow can be used on 1.5mL tubes. Made of polyolefin and acrylic adhesive, it is chemical and temperature resistant from -196-150 deg.C. It comes in 2380 a pack.