Learn More
Andwin Scientific CRYOBABIES YL 33X13 1700/PK
SDP
Catalog No. 50727676
Encompass_Preferred
The 1.28" L x 0.5" W Cryo-Babies labels can withstand most organic solvents, caustic agents, temperatures from -196C to 100C. This product can still be utilized while under oven, autoclaving, or extreme freezer temperatures. The labels can also be used on several surfaces like plastics, glass, and metals without cracking, peeling, or degrading. This product is sold in the color of yellow.
Catalog No. 50-727-676 Supplier Andwin Scientific Supplier No. 77560Y
The 1.28" L x 0.5" W Cryo-Babies labels can withstand most organic solvents, caustic agents, temperatures from -196C to 100C. This product can still be utilized while under oven, autoclaving, or extreme freezer temperatures. The labels can also be used on several surfaces like plastics, glass, and metals without cracking, peeling, or degrading. This product is sold in the color of yellow.