Learn More
Research Products International Corp Cryo-Babies, Laser, 1.5ml, RNBW, 1700/pk
SDP
Catalog No. 502229622
Encompass
Cryo-Babies are designed to offer superior temperature and chemical resistance. Durable labels withstand cryogenic liquid nitrogen storage, boiling water baths, organic solvents, caustic agents and other laboratory chemicals. Cryo-Babies fit cryogenic vials and 1.5 - 2.0 ml micro-tubes. Chemically inert poly vinyl labels adhere to all polypropylene and polyethylene surfaces without cracking, peeling or degrading and accept any marking instrument.
Rainbow pack includes blue, green, orange, red and yellow, four sheets of each.
Catalog No. 50-222-9622 Supplier Research Products International Corp Supplier No. 247127RNBW
Cryo-Babies are designed to offer superior temperature and chemical resistance. Durable labels withstand cryogenic liquid nitrogen storage, boiling water baths, organic solvents, caustic agents and other laboratory chemicals. Cryo-Babies fit cryogenic vials and 1.5 - 2.0 ml micro-tubes. Chemically inert poly vinyl labels adhere to all polypropylene and polyethylene surfaces without cracking, peeling or degrading and accept any marking instrument.
Rainbow pack includes blue, green, orange, red and yellow, four sheets of each.