Learn More
LabScientific Cardboard cell dividers for 64 cells (8x8 format). 12 pack. Fit in Cryovial Boxes. Fits all 2" and 3" cardboard and metal boxes.
Catalog No. NC0639899
Encompass
64 Cardboard Cell Divider (8x8), 12/PK Cell Partitions That Fit In Cryovial Boxes. Fits all 2" and 3" cardboard and metal boxes. Exterior dimensions: 4 7/8 × 4 7/8 × 1 5/16 inch (123.4×123.4×33 mm), Cell opening: 0.57 inch (14.6mm)
Catalog No. NC0639899 Supplier LabScientific Supplier No. CBL64
64 Cardboard Cell Divider (8x8), 12/PK Cell Partitions That Fit In Cryovial Boxes. Fits all 2" and 3" cardboard and metal boxes. Exterior dimensions: 4 7/8 × 4 7/8 × 1 5/16 inch (123.4×123.4×33 mm), Cell opening: 0.57 inch (14.6mm)