Learn More
Azenta US Inc CRYOBOX, HINGED PINK 5/PK
Catalog No. 501311443
Encompass_Preferred
TruCool Hinged CryoBoxes Patented hinged lid offers convenience and archival integrity, ensuring markings and vials remain in sync. Lid stays attached to base minimizing risk of separation and lid contamination. Lid is easy to open when frozen. Available in 9x9,10x10, and vapor phase LN2 compatible formats. Plastic 81-place grid has adjustable slats to accommodate multiple vial types. 2-inch box holds 1.0 mL or 2.0 mL cryogenic vials and microfuge tubes. 3.5-inch box holds 3.0 mL to 5.0 mL cryogenic vials. pink 5 pack Dimensions (L x W x H) 13.3 x 13.3 x 5.1 cm 5.25 x 5.25 x 2.0 in
Catalog No. 50-131-1443 Supplier Azenta US Inc Supplier No. BCS206PK
TruCool Hinged CryoBoxes Patented hinged lid offers convenience and archival integrity, ensuring markings and vials remain in sync. Lid stays attached to base minimizing risk of separation and lid contamination. Lid is easy to open when frozen. Available in 9x9,10x10, and vapor phase LN2 compatible formats. Plastic 81-place grid has adjustable slats to accommodate multiple vial types. 2-inch box holds 1.0 mL or 2.0 mL cryogenic vials and microfuge tubes. 3.5-inch box holds 3.0 mL to 5.0 mL cryogenic vials. pink 5 pack Dimensions (L x W x H) 13.3 x 13.3 x 5.1 cm 5.25 x 5.25 x 2.0 in