Learn More
Cole-Parmer Argos Technologies PolarSafe Cardboard Freezer Box Divider, 5-3/8" x 5-3/8" x 2-3/4", 36-Place, for 15 mL Tubes; 12/pk
Catalog No. NC1652002 Shop All Cole-Parmer Products
Encompass_Preferred
Designed to hold tubes with a max outer diameter of 22 mm. For use in mechanical freezers or liquid nitrogen tanks to 196C.
Catalog No. NC1652002 Supplier Cole-Parmer Supplier No. 0675579
Designed to hold tubes with a max outer diameter of 22 mm. For use in mechanical freezers or liquid nitrogen tanks to 196C.