Learn More
Mitegen CryoCane Magnetic Holder for CryoVials, 5-Vial Capacity, For Liquid Nitrogen & Bulk Storage Tanks, 12 Canes per Pack
SDP
Catalog No. NC1769881
Encompass
The CryoCane Magnetic Holder is engineered to firmly secure magnetic CryoVials (SKUs CV-1-50, CV-1-100, CV-1-200, CV-1-500), optimizing them for efficient liquid nitrogen (LN2) storage within dewars and bulk storage tanks. This innovative holder ensures that valuable samples remain safely in place, preventing accidental loss or disorganization in cryogenic environments. Each CryoCane is designed to hold 5 CryoVials, providing a compact and manageable solution for organizing your cryo-samples. Its robust construction ensures dependable performance in extreme cold. Ideal for laboratories managing extensive cryogenic sample collections, these holders streamline inventory and retrieval processes, contributing to a more organized and safer workspace. The product is supplied as a convenient 12-pack, offering ample capacity for laboratory needs.
Catalog No. NC1769881 Supplier Mitegen Supplier No. MCC301
The CryoCane Magnetic Holder is engineered to firmly secure magnetic CryoVials (SKUs CV-1-50, CV-1-100, CV-1-200, CV-1-500), optimizing them for efficient liquid nitrogen (LN2) storage within dewars and bulk storage tanks. This innovative holder ensures that valuable samples remain safely in place, preventing accidental loss or disorganization in cryogenic environments. Each CryoCane is designed to hold 5 CryoVials, providing a compact and manageable solution for organizing your cryo-samples. Its robust construction ensures dependable performance in extreme cold. Ideal for laboratories managing extensive cryogenic sample collections, these holders streamline inventory and retrieval processes, contributing to a more organized and safer workspace. The product is supplied as a convenient 12-pack, offering ample capacity for laboratory needs.