Learn More
Molecular Dimensions CRYOCANE CODERS (100)
SDP
Catalog No. 501038572
Encompass_Preferred
CryoCane Coders (100) White Rectangular Aluminium tag. Useful colored tabs to keep track of CryoCanes. Available in yellow only. Remain firmly in place during storage in liquid-nitrogen freezers.
Catalog No. 50-103-8572 Supplier Molecular Dimensions Supplier No. MD731
CryoCane Coders (100) White Rectangular Aluminium tag. Useful colored tabs to keep track of CryoCanes. Available in yellow only. Remain firmly in place during storage in liquid-nitrogen freezers.