Learn More
Key Scientific Products CryoCare, Organism Preservative System, Yellow Beads, 50 Vials per Box
SDP
Catalog No. NC9351220
Encompass_Preferred
CryoCare organism preservative system lowers the cost of stock cultures by stretching each expensive culture into 15 or more which meet NCCLS standards. Vial contains ceramic beads in broth base. Includes 50 vials per box with yellow beads.
Catalog No. NC9351220 Supplier Key Scientific Products Supplier No. KS70YE50
CryoCare organism preservative system lowers the cost of stock cultures by stretching each expensive culture into 15 or more which meet NCCLS standards. Vial contains ceramic beads in broth base. Includes 50 vials per box with yellow beads.