Learn More
Key Scientific Products CryoCare Cooked Meat, CryoCare Organism Preservative System, 50 per Box
SDP
Catalog No. NC9943683
Encompass_Preferred
CryoCare lowers the cost of maintaining stock cultures while simplifying freezing and recovery. Cooked meat is especially useful freezing for Streptococci spp and other fastidious organisms.
Catalog No. NC9943683 Supplier Key Scientific Products Supplier No. KSCM50
CryoCare lowers the cost of maintaining stock cultures while simplifying freezing and recovery. Cooked meat is especially useful freezing for Streptococci spp and other fastidious organisms.