Learn More
Key Scientific Products CryoCare Main System, Organism Preservative System, Green Beads, 50 Vials per Box
SDP
Catalog No. NC9841817
Encompass_Preferred
CryoCare organism preservative system lowers the cost of stock cultures by stretching each expensive culture into 15 or more which meet NCCLS standards. Vial contains ceramic beads in broth base. Includes 50 vials per box with green beads.
Catalog No. NC9841817 Supplier Key Scientific Products Supplier No. KS70GR50
CryoCare organism preservative system lowers the cost of stock cultures by stretching each expensive culture into 15 or more which meet NCCLS standards. Vial contains ceramic beads in broth base. Includes 50 vials per box with green beads.