Learn More
Key Scientific Products CryoCare without Beads, CryoCare Organism Preservative System, 50 Vials per Box
SDP
Catalog No. NC9944367
Encompass_Preferred
CryoCare organism preservative system lowers the cost of stock cultures by stretching each expensive culture into 15 or more which meet NCCLS standards. Vial contains a broth base only. Includes 50 vials per box.
Catalog No. NC9944367 Supplier Key Scientific Products Supplier No. KS70X
CryoCare organism preservative system lowers the cost of stock cultures by stretching each expensive culture into 15 or more which meet NCCLS standards. Vial contains a broth base only. Includes 50 vials per box.