Learn More
Thermo Scientific™ CryoClaw
GSA_VA
Catalog No. 1167595 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
All CryoPlus Models
Catalog No. For Use With (Equipment)
11-675-95 All CryoPlus Models
1 options
Catalog No. 11-675-95 Supplier Thermo Scientific™ Supplier No. AY509X18
  • $613.00
    $444.65 / Each of 1
    Save $168.35
    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart
Retrieve vials, ampoules and cryoboxes from cryogenic vessels using the Thermo Scientific™ CryoClaw.
  • Rubber suction cups retrieve ampules or boxes that fall from racks or cans into cryogenic vessels
  • Can also be used to reach overhead items or to safely move radioisotopes without touching containers
  • Length 41in. (104cm)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description CryoClaw Ampule Remover
Type CryoClaw
For Use With (Equipment) All CryoPlus Models
Holds Vials