Learn More
Electron Microscopy Sciences CryoClear™ Cryogenic Vials, Size: 2 mL. Self-Standing: Yes. Bottom: Round., 50/PK
SDP
Supplier:  Electron Microscopy Sciences 6180305
Encompass_Preferred
Cryogenic Vials are long-term protection of samples. They feature screw caps of polyethylene (PE) co-molded with a layer of thermoplastic elastomer (TPE), which provides a superior seal that helps eliminate the risk of sample loss or contamination associated with he use of o-rings vials. The vials has a slim profile cap that provides for a better fit into the racks and boxes, with a star rocket for automated capping and decapping. Each vial has a unique barcode for automated data collection. These Cryogenic caps and vials are autoclavable. Produced with medical grade materials, human DNA/Rnase/Dnase, pyrogen and ATP free. Along with tamper evident packaging. Size: 2 mL. Self-Standing: Yes . Bottom: Round .
Catalog No. 50-209-0637