Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific CryoClear Cryogenic Vials with External Threads

Catalog No. 21402968 Shop All Globe Scientific Products
Change view
Click to view available options
Color:
Blue
Green
Purple
Red
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-402-968 Yellow
21-402-963 Blue
21-402-964 Green
21-402-965 Red
21-402-966 Purple
21-402-967 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 21-402-968 Supplier Globe Scientific Supplier No. 3012Y
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Innovation for the long-term protection of your precious samples, designed specifically for use with automated capping/decapping equipment

  • Globe Scientific CryoClear™ vials feature an innovative polyethylene (PE) screw cap that is co-molded with a layer of thermoplastic elastomer (TPE)
  • This special manufacturing process provides the leak-proof benefits required, while eliminating the risk of contamination and sample loss associated with the use of separate o-rings
  • The best and most leak-resistant cryogenic vials available for the long-term storage of samples at temperatures as low as -196°C
  • Lot certified to be human DNA free, RNase free, DNase free, pyrogen free and ATP free
  • Non-pyrogenic, non-cytotoxic, non-mutagenic, heavy-metals free and BSE/TSE free
  • Manufactured, assembled and packaged in an ISO 7 (Class 10000) cleanroom
  • Slim profile cap design ensures better fit in racks and boxes
  • Unique barcode printed on each vial for automated data collection
  • White writing surface for specimen identification
  • Printed graduations for accurate measurements
  • Vials and caps are autoclavable
  • Produced from medical grade raw materials that will not discolor after re-sterilization
  • Self-standing tubes interlock with Globe workstation racks

Specifications

Color Yellow
Material Polypropylene
Sterility Sterile
Volume (Metric) 2.0 mL
Closure Type Screw Cap
Marking Spot Yes
Autoclavable Autoclavable
Preservative None
Product Type Cryogenic Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.