Learn More
BioSpec Products Cryo-Cup Grinder
SDP
Catalog No. NC1079035
Encompass_Preferred
The Cryo-Cup Grinder consists of a stainless steel, semicircular bowl (or mortar) imbedded in a double walled, insulating cup. Up to 5 grams of hard frozen tissue is placed in the mortar precooled to liq N2 temperature. Using the special hand-held pestle made of non-heat conducting plastic, the tissue is quickly reduced to a powder. Two different sized pestles are provided for a variety of applications.
Catalog No. NC1079035 Supplier BioSpec Products Supplier No. 206
The Cryo-Cup Grinder consists of a stainless steel, semicircular bowl (or mortar) imbedded in a double walled, insulating cup. Up to 5 grams of hard frozen tissue is placed in the mortar precooled to liq N2 temperature. Using the special hand-held pestle made of non-heat conducting plastic, the tissue is quickly reduced to a powder. Two different sized pestles are provided for a variety of applications.