Learn More
Research Products International Corp CryoELITE 2.0ml Cryogenic Vials, 12 x 49mm, Sterile, Yellow, 100 per Case
SDP
Catalog No. 501367878
Encompass
CryoELITE Cryogenic Vials offer the ultimate sample integrity for storing, collecting and transporting valuable tissue. Manufactured from low binding polypropylene that meets USP Class VI classification. Each lot is certified free of RNase, DNase, Endotoxins and Pyrogens. Vials feature external threads, writing surface and Loctagon Vial Skirt for stability during free standing position. Vials also lock into CryoELITE Benchmate Rack to facilitate convenient one hand operation.
Available in Blue, Green, Pink, Red, White and Yellow
Catalog No. 50-136-7878 Supplier Research Products International Corp Supplier No. 212320Y
CryoELITE Cryogenic Vials offer the ultimate sample integrity for storing, collecting and transporting valuable tissue. Manufactured from low binding polypropylene that meets USP Class VI classification. Each lot is certified free of RNase, DNase, Endotoxins and Pyrogens. Vials feature external threads, writing surface and Loctagon Vial Skirt for stability during free standing position. Vials also lock into CryoELITE Benchmate Rack to facilitate convenient one hand operation.
Available in Blue, Green, Pink, Red, White and Yellow