Learn More
Research Products International Corp CryoELITE 5ml Tissue Vials, Sterile, 25 per Package
SDP
Catalog No. 501367691
Encompass
CryoELITE Tissue Vial provides the ultimate sample integrity for storing and transporting tissue. Manufactured from low binding polypropylene with each lot certified free of RNase, DNase, Endotixins and Pyrogens. Features include a wide mouth opening with external threads; flat bottom and a stippled exterior surface for easier handling.
Catalog No. 50-136-7691 Supplier Research Products International Corp Supplier No. 212310
CryoELITE Tissue Vial provides the ultimate sample integrity for storing and transporting tissue. Manufactured from low binding polypropylene with each lot certified free of RNase, DNase, Endotixins and Pyrogens. Features include a wide mouth opening with external threads; flat bottom and a stippled exterior surface for easier handling.