Learn More
Research Products International Corp CryoELITE Storage Box, Holds 25 Tissue Vials, 5 Boxes per Case
SDP
Catalog No. 501367692
Encompass
CryoELITE Storage Box is designed for storing CryoELITE Tissue Vials at low cryogenic temperatures. All components are finished in a white color and are water repellent for longer durability. Twenty five separate compartments are individually numbered for quick indexing. Two boxes can be stacked together to fit standard 2" high freezer racks. Storage boxes withstand temperatures to -196°C.
Catalog No. 50-136-7692 Supplier Research Products International Corp Supplier No. 212311
CryoELITE Storage Box is designed for storing CryoELITE Tissue Vials at low cryogenic temperatures. All components are finished in a white color and are water repellent for longer durability. Twenty five separate compartments are individually numbered for quick indexing. Two boxes can be stacked together to fit standard 2" high freezer racks. Storage boxes withstand temperatures to -196°C.